Lipase, MP Biomedicals
Supplier: MP Biomedicals
0210218990
ICNA0210218990EA 660 SGD
ICNA0210218990
Lipase, MP Biomedicals
Enzymes
One USP unit of lipase activity is contained in the amount of pancreatin that liberates 1,0 µEq of acid per minute at a pH of 9,0 at 37 °C under the conditions of the assay for lipase activity.
  • Partly soluble in water and insoluble in alcohol
Lipases are a subclass of the esterases. It catalyses the hydrolysis of emulsified esters of glycerol and long chain fatty acids.
Order Now
SPECIFICATIONS
Enzyme name Lipase
Environmentally Preferable
Source
Species
CAS No. 9001-62-1