Melatonin, off-white powder

Supplier: MP Biomedicals

02102254.1 0210225401 0210225405 0210225410
ICNA02102254.1EA 85.5 SGD
ICNA02102254.1 ICNA0210225401 ICNA0210225405 ICNA0210225410
Melatonin, off-white powder
Melatonine

Melatonin is a hormone of the pineal gland.


  • Light sensitive
  • Subject to oxidation


Hormone; mediates photoperiodicity in mammals; inhibits cerebellar nitric oxide synthetase; peroxynitrite scavenger. Melatonin has complex effects on apoptotic pathways, inhibiting apoptosis in immune cells and neurons but enhancing apoptotic cell death of cancer cells. Inhibits proliferation/metastasis of breast cancer cells by inhibiting estrogen receptor action.


Soluble in methanol, ethanol (50 mg/ml - clear to slightly hazy, colorless to faint yellow solution); very slightly soluble in 0,1 N HCl (0,8 mg/ml), 0,1 N NaOH (0,2 mg/ml) or water (0,1 mg/ml).


Caution: Research Use Only (RUO).

Formula: C₁₃H₁₆N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00005655
CAS Number: 73-31-4

Order Now

Specification Test Results

Identity Test Passes
Melting Point 114 - 124 °C
FTIR Conforms to Standard
UV/Vis: λmax (methanol) 222 - 225, 276 - 279 nm
Moisture ≤1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR