L-Methionine

Supplier: MP Biomedicals

02102291.1 02102291.5 0210229101 02102291.3
ICNA02102291.1EA 150 SGD
ICNA02102291.1 ICNA02102291.5 ICNA0210229101 ICNA02102291.3
L-Methionine
L-Methionine
Formula: C₅H₁₁NO₂S
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 136 °C
Density: 1,373 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR