L(-)-Tryptophan 99, powder

Supplier: MP Biomedicals

Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

0210315190 0210315125 0210315105 0210315180 0210315191
ICNA0210315190EA 961 SGD
ICNA0210315190 ICNA0210315125 ICNA0210315105 ICNA0210315180 ICNA0210315191
L(-)-Tryptophan 99, powder
L(-)-Tryptophan

L-Tryptophan is an amino acid precursor of serotonin and melatonin.


Chloride ≤0,05%
sulphate ≤0,03%
Iron ≤0,003%


It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.


It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.


Soluble in 1N HCl (10 mg/ml - clear, colourless solution).

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 290
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Order Now

Specification Test Results

Synonyms L-2-amino-3-indolepropionic acid; Trp; W; (S)-2-Amino-3-(3-indolyl)propionic acid
Form White Powder
Storage Store at room temperature (15 to 30 °C)
Heavy Metals ≤0,0015%
Moisture Content ≤0,3%
Optical Rotation −30° ±10° (c=1, water)
pH 5,5 - 7,0 (1% aq soln)
Residue on Ignition ≤0,1%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR