Xanthine sodium salt 98, pellets

Supplier: MP Biomedicals
Danger

Synonyms: 2,6-Dihydroxypurine

0210329125 0210329105
ICNA0210329125EA 0 SGD
ICNA0210329125 ICNA0210329105
Xanthine sodium salt 98, pellets
Xanthine sodium salt

The methylated xanthine compounds caffeine, theobromine, and theophylline and their derivatives are used in medicine for their bronchodilator effects.


UV Visible Absorbance λ max (0.01 N HCl)=265 ± 5nm

Formula: C₅H₃N₄NaO₂
MW: 174.095 g/mol
MDL Number: MFCD00069699
CAS Number: 1196-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR