D-(+)-Xylose, white powder

Supplier: MP Biomedicals

0210330080 0210330090
ICNA0210330080EA 77 SGD
ICNA0210330080 ICNA0210330090
D-(+)-Xylose, white powder
D(+)-Xylose

Xylose is the main building block for hemicellulose, the acid-catalysed degradation of hemicellulose gives furfural, a specialty solvent in industry and a precursor to synthetic polymers. In animal medicine, xylose is used to test for malabsorption by administration in water to the patient after fasting.


Soluble in water (clear,colorless solution), pyridine, and hot alcohol.


Caution: Research Use Only (RUO).

Formula: C₅H₁₀O₅
Melting Pt: 155…157 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Optical Rotation +18° ± 5° (c=4, H2O, NH4OH)
Loss on Drying ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR