N-Octyl-β-D-glucopyranoside 98, powder_white

Supplier: MP Biomedicals
Warning

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

02150054.2 02150054.5 0215005405 0215005425 0215005401 0215005410
ICNA02150054.2EA 132 SGD
ICNA02150054.2 ICNA02150054.5 ICNA0215005405 ICNA0215005425 ICNA0215005401 ICNA0215005410
N-Octyl-β-D-glucopyranoside 98, powder_white
N-Octyl-β-D-glucopyranoside

n-Octyl-β-D-Glucopyranoside is a non-ionic detergent for membrane protein.


n-Octyl-glucoside is a non-ionic alkyl-glucoside detergent commonly used in the solubilization of membrane proteins and phospholipids. n-Octyl-glucoside has been used at a molar dependant ratios for the solubilization of phosphatidylcholine bilayers. n-Octyl-glucoside has been used for protein crystallization, as well as for enhancement of oligosaccharide cleavage from glycoproteins using endoglycosidase F.


Soluble in water (10 mg/ml - clear colourless solution). 20-25 mM Detergents with high CMC values are generally easy to remove by dilution; detergents with low CMC values are advantageous for separations on the basis of molecular weight. As a general rule, detergents should be used at their CMC and at a detergent-to-protein weight ratio of approximately ten.

Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 89…100 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00063288
CAS Number: 29836-26-8
EINECS: 249-887-8

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Optical Rotation (c=5, water) -31 ± 3°

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR