Ammonium carbonate, white powder

Supplier: MP Biomedicals

Synonyms: Salt of hartshorn

0215010880
ICNA0215010880EA 117 SGD
ICNA0215010880
Ammonium carbonate, white powder
Ammonium carbonate

Ammonium carbonate is used in the washing and defatting of wools, the manufacture of rubber products, in fire extinguishers, and the separation of cacao constituents.


A high-throughput parallel liquid chromatography/ultraviolet/mass spectrometry (LC/UV/MS) technique for compound screening that incorporates ammonium carbonate has been published. Ammonium carbonate can be used in the isolation of cell surface proteins from Candida albicans. It also finds application in the comparative peptide analysis of equine, bovine, and canine cytochrome c by capillary electrophoresis/mass spectrometry. The preparation of glycoconjugates via Kochetkov aminations in saturated aqueous ammonium carbonate has been reported.


Soluble in 4 parts water, decomposed by hot water.


Caution: Research Use Only (RUO).

Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
Melting Pt: 58 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 10361-29-2
Merck Index: 13,00508

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR