Ammonium sulphate ≥99%, white crystalline powder, Enzyme Grade

Supplier: MP Biomedicals
Warning

0215037390 0215037380 0215037391
ICNA0215037390EA 161 SGD
ICNA0215037390 ICNA0215037380 ICNA0215037391
Ammonium sulphate ≥99%, white crystalline powder, Enzyme Grade
Ammonium sulphate

Ammonium Sulfate may be used for the precipitation or fractionation of proteins or for purification of antibodies.


Useful for crystallographic analysis of nucleic acids and proteins. The use of ammonium sulfate in long PCR buffer, in PCR lysis solution, and in second strand cDNA synthesis has been reported.


Soluble freely in water; insoluble in alcohol, acetone.

Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1,78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Purity ≥99%
Heavy Metals ≤0,5 ppm (as Pb)
pH 5 - 6 (5% aq soln)
Protease, DNase, RNase Not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR