L-Aspartic acid magnesium salt, powder

Supplier: MP Biomedicals

0215040691 0215040625
ICNA0215040691EA 1860 SGD
ICNA0215040691 ICNA0215040625
L-Aspartic acid magnesium salt, powder
L-Aspartic acid magnesium salt

Freely soluble in water.

Formula: C₈H₁₂MgN₂O₈
CAS Number: 18962-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR