Cesium chloride 99,999, powder_white, ultrapure

Supplier: MP Biomedicals

Synonyms: Cesium Chloride

0215058905 0215058925 0215058980 0215058990 0215058991
ICNA0215058905EA 491 SGD
ICNA0215058905 ICNA0215058925 ICNA0215058980 ICNA0215058990 ICNA0215058991
Cesium chloride 99,999, powder_white, ultrapure
Cesium chloride

Cesium chloride is used for the preparation of electrically conducting glasses. It is also used to make solutions for the separation of RNA from DNA by density gradient centrifugation.


  • A260 of 50% solution < 0.02
  • Solutions are clear and colorless
  • Especially suited for critical density gradient techniques.


pH: 5.5 - 6.5 (1% aq soln)


Very soluble in water. Soluble in ethanol.

Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303
Melting Pt: 646
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
Merck Index: 13,02021

Order Now

Specification Test Results

Purity 99.999%
pH 5,5 - 6,5 (1% aq soln)
Refractive Index 1,6418 (Lit.)
Barium ≤1 ppm
Magnesium ≤0,5 ppm
Potassium ≤2 ppm
Iron ≤1 ppm
Calcium ≤1 ppm
OD260 nm (50% aq soln) <0.02

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR