Chitosan Crustacean chitin, powder_off-white

Supplier: MP Biomedicals

Synonyms: Deacetylated chitin, Poly(D-glucosamine)

0215059783 0215059710 0215059725 0215059780
ICNA0215059783EA 840 SGD
ICNA0215059783 ICNA0215059710 ICNA0215059725 ICNA0215059780
Chitosan Crustacean chitin, powder_off-white
Chitosan

Chitosan is a cationic marine polymer derived form the shells of crustaceans (e.g., crabs). MP's chitosan is a partially de-acetylated chitin formed by the reaction of chitin with concentrated alkali. Chitin is a high molecular weight linear polymer of anhydro-N-acetyl-glucosamine. Chitin occurs in nature as part of the protective coating of some invertebrates.


Chitosan forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films. It is biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.


Caution: Research Use Only (RUO).

Density: 0.15…0.3 g/cm³ (bulk density)
Storage Temperature: Ambient
MDL Number: MFCD00161512
CAS Number: 9012-76-4

Order Now

Specification Test Results

Identity Test Passes
Source Crustacean Chitin
Moisture ≤10%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR