Collagenase, MP Biomedicals
Supplier: MP Biomedicals
0215070401
ICNA0215070401EA 1600 SGD
ICNA0215070401
Collagenase, MP Biomedicals
Enzymes
For typical assay, dissolve enzyme at a concentration of 1 mg/ml in 0,05 M TES with 0,36 mM calcium chloride, pH 7,5. Dilutions run are 1/10 and 1/20 in the above buffer.
One unit liberates one µmole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7,5.
Activators: Ca²⁺ is required; Inhibitors: Metal chelating agents such as cysteine, EDTA or o-phenanthroline but not DFP. It is also inhibited by a2-macroglobulin.