Collagenase, MP Biomedicals
Supplier: MP Biomedicals
0215070501
ICNA0215070501EA 1610 SGD
ICNA0215070501
Collagenase, MP Biomedicals
Enzymes
Collagenase can be solubilised in calcium-free solutions such as Hank's Balanced Salts or Earles Balanced Salt Solution.
Unit definition: One unit liberates one µmole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7,5.