3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) synthetic, powder

Supplier: MP Biomedicals

Synonyms: 3,3',4,4'-Biphenyltetramine tetrahydrochloride, DAB HCl

0215082510 0215082501 0215082505
ICNA0215082510EA 1130 SGD
ICNA0215082510 ICNA0215082501 ICNA0215082505
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) synthetic, powder
3,3'-Diaminobenzidine tetrahydrochloride

Easily soluble in water. The aqueous solution is unstable and can be used only for a few days even if it is kept in a refrigerator. 1% aqueous solution is dark and turns reddish when exposed to light.


Colour changes to grey or grey brown when exposed to the direct sun light.

Formula: C₁₂H₁₈Cl₄N₄
MW: 360.11 g/mol
MDL Number: MFCD00012969
CAS Number: 7411-49-6
EINECS: 231-018-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR