Dihydromyrcenol, liquid

Supplier: MP Biomedicals

0215090680 0215090625
ICNA0215090680EA 386 SGD
ICNA0215090680 ICNA0215090625
Dihydromyrcenol, liquid
Dihydromyrcenol
Formula: C₁₀H₂₀O
CAS Number: 18479-58-8
EINECS: 242-362-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR