Guanidinium hydrochloride 99, powder_crystalline_white Optical Density Grade

Supplier: MP Biomedicals

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

0215121025 0215121080 0215121090 0215121091
ICNA0215121025EA 85.2 SGD
ICNA0215121025 ICNA0215121080 ICNA0215121090 ICNA0215121091
Guanidinium hydrochloride 99, powder_crystalline_white Optical Density Grade
Guanidinium hydrochloride

Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins.


This strong denaturant can solubilise insoluble or denatured proteins such as inclusion bodies and be used for the recovery of periplasmic proteins. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. It is also used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties. It is an inhibitor of RNase.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
MDL Number: MFCD00013026
CAS Number: 50-01-1
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 180 - 190 °C
OD260 (6,0 M aq soln) ≤0,03
Solubility (6,0 M aq soln) C Clear, Colorless Solution

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR