γ-L-Glutamyl-p-nitroanilide hydrochloride, beige powder

Supplier: MP Biomedicals

0215149501 02151495.2 0215149505
ICNA0215149501EA 286 SGD
ICNA0215149501 ICNA02151495.2 ICNA0215149505
γ-L-Glutamyl-p-nitroanilide hydrochloride, beige powder
γ-L-Glutamyl-p-nitroanilide hydrochloride

γ-L-Glutamyl-p-Nitroanilide hydrochloride is suitable for use as a substrate for g-Glutamyl transpeptidase.


Soluble in water (5 mg/ml - clear, light yellow solution) or 0,1 N HCl (1% w/v - clear, yellowish solution); soluble in methanol (100 mg/ml with slight heat and constant stirring).


Caution: Research Use Only (RUO).

Formula: C₁₁H₁₄ClN₃O₅
MDL Number: MFCD00039054
CAS Number: 67953-08-6

Order Now

Specification Test Results

Identity Test Passes
Optical Rotation (c=1, 0.1 N HCl) +30° ± 5°
UV/Vis: λmax (water) 317 ± 5 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR