3-Methylxanthine, powder_white

Supplier: MP Biomedicals
Warning

Synonyms: 3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine

0215168680 0215168683
ICNA0215168680EA 129 SGD
ICNA0215168680 ICNA0215168683
3-Methylxanthine, powder_white
3-Methylxanthine

3-Methylxanthine, a metabolite of theophylline, was quantitated in rat plasma by a sensitive LC-MS/MS method.


Freely soluble in alkali hydroxides and in mineral acid; soluble in dilute aqueous base or ethanol; slightly soluble in hot water; practically insoluble in organic solvent

Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005580
CAS Number: 1076-22-8
EINECS: 214-058-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR