Neuraminidase (from Bacteria), MP Biomedicals

Supplier: MP Biomedicals

0215173805
ICNA0215173805EA 845 SGD
ICNA0215173805
Neuraminidase (from Bacteria), MP Biomedicals
Enzymes

Neuraminidase of non-pathogenic streptococci, is prepared from the culture filtrate by Ammonium Sulfate precipitation and Ion-exchange chromatography.


  • Enzyme Commission Number: E.C.3.2.1.18
  • Activators: Mg2+, Co2+, Ca2+, and K+ (10mM), these activators accelerate activity 10 to 20%
  • Inhibitors: Ag+, Hg+, Cu2+, and Potassium Permanganate (10mM)
  • Isoelectric point (pI): 7,7 ±0,1


Unit definition: One unit causes the hydrolysis of one µmole of sialyllactose per minute at pH 6,5, 37 °C.

Order Now

SPECIFICATIONS

Enzyme name Neuraminidase
Environmentally Preferable
Enzyme synonyms Acylneuramyl hydrolase
Source Bacteria
Species
CAS No. 9001-67-6
Molecular weight 85 kDa
Form White lyophilized powder
Storage conditions 2...8 °C
Enzyme activity ≥50 u/mg solid
pH 6,0 - 6,5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR