Cellulase (from Trichoderma virde), MP Biomedicals
Supplier: MP Biomedicals
0215233705
ICNA0215233705EA 7860 SGD
ICNA0215233705
Cellulase (from Trichoderma virde), MP Biomedicals
Enzymes
Readily soluble in water.
One unit will liberate one µmole of glucose from cellulose in 1 hr at pH 5 and 37 °C. ; CMC Unit Definition: 100 units is sufficient to produce reducing sugar equivalent to 1 mg of glucose per minute at pH 5,0 and 40 °C.