Lanthanum(III) chloride heptahydrate, clear to white crystalline powder ACS

Supplier: MP Biomedicals
Warning

0215253480 0215253425 0215253490
ICNA0215253480EA 356 SGD
ICNA0215253480 ICNA0215253425 ICNA0215253490
Lanthanum(III) chloride heptahydrate, clear to white crystalline powder ACS
Lanthanum (III) chloride heptahydrate

Calcium channel blocker.


  • Calcium channel blocker


Lanthanum chloride is used to precipitate phosphate from solutions. Lanthanum chloride is also used in biochemical research to block the activity of divalent cation channels, mainly calcium channels.

Formula: LaCl₃·7H₂O
Melting Pt: 91 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0

Order Now

Specification Test Results

Assay 64.5 - 70.0% LaCl3
Insoluble Matter ≤0.01%
Calcium (Ca) ≤0.001%
Magnesium (Mg) ≤10ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR