Basic blue 3, dark green powder C.I. 51004

Supplier: MP Biomedicals

0215262910
ICNA0215262910EA 133 SGD
ICNA0215262910
Basic blue 3, dark green powder C.I. 51004
Basic blue 3

Soluble in water, EGME, ethanol.


Soluble in water, EGME, ethanol.


Caution: Research Use Only (RUO).

Formula: C₂₀H₂₆N₃OCl
Storage Temperature: Ambient
MDL Number: MFCD00011928
CAS Number: 33203-82-6

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: λmax (water) Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR