N-t-Boc-Phe-Leu-Phe-Leu-Phe, MP Biomedicals
Supplier: MP Biomedicals
0215276005
ICNA0215276005EA 501 SGD
ICNA0215276005
N-t-Boc-Phe-Leu-Phe-Leu-Phe, MP Biomedicals
Proteins and Peptides
Soluble in 1:1 CHCl₃:methanol (50 mg/ml), soluble in DMF ((25 mg/ml).
N-Boc-Phe-Leu-Phe-Leu-Phe(Boc-PLPLP) is used extensively in Formyl Peptide Receptor(FPR) research. In regard to neutrophils, it was found that treatment of mice with Boc-PLPLP results in 40% reduction in neutrophil counts recovered in bronchoalveolar lavage fluid, compared with infected mice receiving placebo.