Sodium alginate (from Macrocystis pyrifera (kelp)), off-white powder, low viscosity

Supplier: MP Biomedicals
Warning

0215472580 0215472583 0215472590 0215472591
ICNA0215472580EA 79.7 SGD
ICNA0215472580 ICNA0215472583 ICNA0215472590 ICNA0215472591
Sodium alginate (from Macrocystis pyrifera (kelp)), off-white powder, low viscosity
Sodium alginate

A gelling polysaccharide extracted from giant brown seaweed. It is a straight chain, colloidal, polyuronic acid composed mainly of anhydro-b-D-mannuronic acid residues linked in the 1---->4 position. It has been used in the flocculation of solids in water treatment; as sizing agent; thickener; emulsion stabilizer; and a suspending agent.
Alginate, a colloidal polyuronic acid structural molecule capable of gelation, is used in the preparation of colloidal biodegradable structures such as gels, biofilms, beads, nanoparticles, and microcapsules suitable for applications that range from gel based separation technologies to drug delivery and cell preservation.


  • Low Viscosity
  • Presentation: Off-White Powder
  • Soluble in water, forming a viscous, colloidal solution.
  • Insoluble in alcohol and in hydro-alcoholic solutions in which the alcohol content is >30% w/w.
  • Insoluble in chloroform, ether, in aqueous acid solutions when the pH is below 3.
  • Storage temperature: Store at Room Temperature (15-30 °C)

Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now

Specification Test Results

Identity Test Passes
Source Macrocystis pyrifera (Kelp)
Viscosity (1% aq soln) ≤300 cps


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR