2-Bromethylamine hydrobromide 98,0, powder_crystalline_white

Supplier: MP Biomedicals
Warning

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

0215487525 0215487580
ICNA0215487525EA 99.1 SGD
ICNA0215487525 ICNA0215487580
2-Bromethylamine hydrobromide 98,0, powder_crystalline_white
2-Bromethylamine hydrobromide

2-Bromoethylamine is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone.


Caution: Research Use Only (RUO).

Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 342…345° F (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR