Melanin

Supplier: MP Biomedicals

0215534391 0215534394 0215534380 0215534383
ICNA0215534391EA 946 SGD
ICNA0215534391 ICNA0215534394 ICNA0215534380 ICNA0215534383
Melanin
Melanin

Soluble in dilute NH3.


Prepared by persulphate oxidation of tyrosine.


Melanin is a natural substance that gives color (pigment) to hair, skin, and the iris of the eye. It is produced by cells in the skin called melanocytes. Melanin also helps protect the skin from the sun. Melanin is an extremely stable molecule, and highly resistant to the digestion by most acids and bases.


Melanin is used to prepare inhibitor in Real Time PCR analysis.

Formula: C₁₈H₁₀N₂O₄
MW: 318.288 g/mol
MDL Number: MFCD00131581
CAS Number: 8049-97-6

Order Now

Specification Test Results

Form Dark Gray Powder
Storage Store at −0 °C.
Source Synthetic

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR