Nitrofurantoin, yellow powder

Supplier: MP Biomedicals

0215588125 0215588180 0215588183
ICNA0215588125EA 134 SGD
ICNA0215588125 ICNA0215588180 ICNA0215588183
Nitrofurantoin, yellow powder
Nitrofurantoin

Nitrofurantoin is an antibactericidal compound that has been historically prepared by the reaction of 1-aminohydantoin sulphate and 5-nitro-2-furaldehyde diacetate.


It shows activity against many gram positive and gram-negative bacteria. Nitrofurantoin is effective against enterococci, staphylococci, streptococci, corney bacteria, many strains of escherichia coli. By contrast, most strains of proteus spp. and pseudomonas aeurginosa are more resistant to this compound. Other microbial species whose susceptibility to nitrofurantoin has been studied include plesiomonas shigelloide, campylobacter, and providencia.


Nitrofurantoin is activated by bacterial flavoproteins (nitrofuran reductase) to actively reduce reactive intermediates that modulate and damage ribosomal proteins or other macromolecules, such as DNA. This inhibits DNA, RNA, protein, and cell wall synthesis which causes cell death. Nitrofurantoin has a low resistance potential that is rapidly metabolised by mammals and is active against both gram-positive and gram-negative bacteria. It is also a pro-oxidant that is cytotoxic due to the generation of intracellular H2O2. It is an inhibitor of glutathione reductase. Nitrofurantoin produces hepatotoxicity caused by the redox cycling of the nitro group and its radical anion which leads to oxidative stress.


Soluble in dimethylformamide (50 mg/ml - clear yellow to yellow-green solution), water, ethanol, acetone, peanut oil, glycerol, polyethylene glycol.

Formula: C₈H₆N₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00003224
CAS Number: 67-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR