Colophony (Rosin gum), chunks

Supplier: MP Biomedicals

Synonyms: Rosin, gum, Rosin, Gum rosin

0215655790 0215655791
ICNA0215655790EA 330 SGD
ICNA0215655790 ICNA0215655791
Colophony (Rosin gum), chunks
Colophony (Rosin gum)

Soluble in alcohol (ethanol: 50 mg/ml), benzene, ether. Insoluble in water. Chloroform: soluble 0,1 g/ml, clear, slightly yellow to deep yellow.


Soluble in alcohol (ethanol: 50 mg/ml), benzene, ether. Insoluble in water. Chloroform: soluble 0,1 g/ml, clear, slightly yellow to deep yellow.

Melting Pt: 70…130 °C (lit.)
Flash Pt: 188
Storage Temperature: Ambient
MDL Number: MFCD00132205
CAS Number: 8050-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR