1-Hexanol, liquid

Supplier: MP Biomedicals

Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane

0215736280
ICNA0215736280EA 0 SGD
ICNA0215736280
1-Hexanol, liquid
1-Hexanol
Formula: H₃C(CH₂)₄CH₂OH
MW: 102.18 g/mol
Boiling Pt: 156
Melting Pt: -45
Flash Pt: 60
MDL Number: MFCD00002982
CAS Number: 111-27-3
UN: 2282
ADR: 3 III
Merck Index: 13,04715
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=5117270d1d57dd4b9cca9ee3d787375106cc1216&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR