20-Hydroxyecdysone 95, powder_white

Supplier: MP Biomedicals

Synonyms: β-Ecdysone

0215743405
ICNA0215743405EA 338 SGD
ICNA0215743405
20-Hydroxyecdysone 95, powder_white
20-Hydroxyecdysone

20-Hydroxyecdysone is the most widely occurring ecdysteroid in both plant and animal species. Controls cell death during metamorphosis of Drosophila melanogaster.


20-Hydroxyecdysone is an ecdysteroid hormone that plays a key role in insect development, cell proliferaton, growth and apoptosis by controlling gene expression involved in molting and metamorphosis. It acts through a heterodimeric receptor comprising the ecdysone receptor and the ultraspiracle proteins (USP).


Soluble in methanol 20 mg/ml; easily soluble in ethanol and soluble in water.

Formula: C₂₇H₄₄O₇
MW: 480.642 g/mol
MDL Number: MFCD00036740
CAS Number: 5289-74-7

Order Now

Specification Test Results

Identity Test Passes
Purity ≥95%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR