β-Escin, white 92 - 95%, powder

Supplier: MP Biomedicals
Warning

Synonyms: beta-Escin, β-Aescin

0215794105 0215794101
ICNA0215794105EA 648 SGD
ICNA0215794105 ICNA0215794101
β-Escin, white 92 - 95%, powder
β-Escin
MW: 1101.2 g/mol
MDL Number: MFCD00076054
CAS Number: 11072-93-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR