Benzocaine (ethyl-4-aminobenzoate), powder_crystalline_white

Supplier: MP Biomedicals
Warning

Synonyms: Ethyl 4-aminobenzoate, Aethoform

0215796790 0215796780
ICNA0215796790EA 1320 SGD
ICNA0215796790 ICNA0215796780
Benzocaine (ethyl-4-aminobenzoate), powder_crystalline_white
Benzocaine (Ethyl 4-aminobenzoate)
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 311
Melting Pt: 88…93 °C
Flash Pt: 112
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5
Merck Index: 13,01087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR