n-Undecane 99, Liquid_clear, colorless

Supplier: MP Biomedicals
Danger

Synonyms: Undecane

0215823180 0215823190
ICNA0215823180EA 122 SGD
ICNA0215823180 ICNA0215823190
n-Undecane 99, Liquid_clear, colorless
n-Undecane

Undecane may be used as an internal standard in gas chromatography when working with other hydrocarbons. Since the boiling point of undecane (196 °C) is well known, it may be used as a comparison for retention times in a gas chromatograph for molecules whose structure has been freshly elucidated.


  • Japan fire code: 4.2

Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
Boiling Pt: 196
Melting Pt: -26.7
Flash Pt: 60
MDL Number: MFCD00008959
CAS Number: 1120-21-4
UN: 2330
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR