Protease inhibitor cocktail, MP Biomedicals
Supplier: MP Biomedicals
0215883701
ICNA0215883701EA 358 SGD
ICNA0215883701
Protease inhibitor cocktail, MP Biomedicals
Cell Culture Supplements and Additives
A broad spectrum protease inhibitor cocktail which covers both endopeptidases and exopeptidases, non-specific and specific. The cocktail consists of four major protease inhibitors such as AEBSF, Sodium EDTA, Leupeptin and Pepstain A.