Kifunensine ≥98%, white powder

Supplier: MP Biomedicals

0215995201
ICNA0215995201EA 471 SGD
ICNA0215995201
Kifunensine ≥98%, white powder
Kifunensine

Kifunensine is an alkaloid compound, originally isolated from the actinomycete Kitasporia kifunensis, that exhibits potent and selective inhibition of class I α-mannosidases. Kifunensine is used to suppress Endoplasmic Reticulum-Associated Degradation (ERAD) via the inhibition of endoplasmic reticulum-associated mannosidase activity.


Selective inhibitor of class I glycoprotein-processing α-mannosidases.


Soluble in water (25 mg/ml).

Formula: C₈H₁₂N₂O₆
MW: 232.192 g/mol
Storage Temperature: Freezer
CAS Number: 109944-15-2

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR