Myrothecium sp. Bilirubin oxidase, MP Biomedicals

Supplier: MP Biomedicals

0219000010 0219000020 0219000080
ICNA0219000010EA 139 SGD
ICNA0219000010 ICNA0219000020 ICNA0219000080
Myrothecium sp. Bilirubin oxidase, MP Biomedicals
Enzymes

  • Lyophilised powder
  • Activity: ≥0,8 u/mg solid
  • Unit definition: one unit is the amount of enzyme that oxidises one micromole of bilirubin per minute at 37 °C, pH 8,4.

Order Now

SPECIFICATIONS

Enzyme name Bilirubin Oxidase
Environmentally Preferable
Source
Species Myrothecium sp.
CAS No. 80619-01-8
Storage conditions +4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR