Myrothecium sp. Bilirubin oxidase, MP Biomedicals
Supplier: MP Biomedicals
0219000010 0219000020 0219000080
ICNA0219000010EA 139 SGD
ICNA0219000010 ICNA0219000020 ICNA0219000080
Myrothecium sp. Bilirubin oxidase, MP Biomedicals
Enzymes
  • Lyophilised powder
  • Activity: ≥0,8 u/mg solid
  • Unit definition: one unit is the amount of enzyme that oxidises one micromole of bilirubin per minute at 37 °C, pH 8,4.