Methylbenzethonium chloride, white powder

Supplier: MP Biomedicals
Danger

0219024010 0219024025
ICNA0219024010EA 871 SGD
ICNA0219024010 ICNA0219024025
Methylbenzethonium chloride, white powder
Methylbenzethonium chloride

Methylbenzethonium Chloride is a quaternary ammonium salt that has a surface action similar to that of other cationic detergents.


It is generally germicidal and bacteriostatic and is most commonly used to rinse infant diapers and bed linen in the prevention of ammonia dermatitis. It has been shown to kill microorganisms, or prevent or inhibit their growth, and thus protect cosmetics and personal care products from spoilage. They also help to distribute or to suspend an insoluble solid in a liquid.


Methylbenzethonium chloride has been used in a study to assess stem cell death-inducing small molecules. It has also been used in a study to investigate the anti-leishmanial activity of paromomycin ointment (Leshcutan) combined with the immunomodulator Imiquimod.


Soluble in water (1 g/20 mL).

Formula: C₂₈H₄₄ClNO₂
Melting Pt: 159…162 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011820
CAS Number: 25155-18-4

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR