Clofibrate

Supplier: MP Biomedicals

Synonyms: Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate

02190342.2 0219034205
ICNA02190342.2EA 112 SGD
ICNA02190342.2 ICNA0219034205
Clofibrate
Clofibrate

Practically insoluble in water. Miscible with ethanol, acetone, chloroform, ether.

Formula: C₁₂H₁₅ClO₃
MW: 242.70 g/mol
MDL Number: MFCD00000615
CAS Number: 637-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR