Isodesmosine ≥99%, yellow solid

Supplier: MP Biomedicals

Synonyms: 2-(4-Amino-4-carboxybutyl)-1-(5-amino-5-carboxypentyl)-3,5-bis(3-amino-3-carboxypropyl)pyridinium chloride, 2-amino-6-[3,5-bis(3-amino-4-hydroxy-4-oxobutyl)-2-(4-amino-5-hydroxy-5-oxopentyl)pyridin-1-ium-1-yl]hexanoic acid, Iso-desmosin

0219137901 0219137910
ICNA0219137901EA 853 SGD
ICNA0219137901 ICNA0219137910
Isodesmosine ≥99%, yellow solid
Isodesmosine

Isodesmosine is prepared according to the method of Starcher, et al. from hydrolysed bovine neck ligament by solvent extraction and chromatography on Dowex 50. It is chromatographically purified hygroscopic crystalline powder.


Caution: Research Use Only (RUO).

Formula: C₂₄H₄₀N₅O₈+
Storage Temperature: Refrigerator
MDL Number: MFCD00211039
CAS Number: 991-01-5

Order Now

Specification Test Results

Identity Test Passes
Source Bovine Neck Ligament
Purity (amino acid analysis) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR