Barium chloride dihydrate ≥99.0%, powder ACS

Supplier: MP Biomedicals
Danger

0219141080 0219141090
ICNA0219141080EA 90.8 SGD
ICNA0219141080 ICNA0219141090
Barium chloride dihydrate ≥99.0%, powder ACS
Barium chloride dihydrate

Soluble in water.

Formula: BaCl₂·2H₂O
MDL Number: MFCD00149154
CAS Number: 10326-27-9
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Form White Granular Powder
Storage Store at Room Temperature (15 - 30 °C)
Assay ≥99.0%
Heavy Metals ≤5 ppm
Insoluble Matter ≤0,005%
Loss on Drying 14,0 - 16,0%
pH 5,2 - 8,2 (5% aq soln)
Oxidizing Substances (as NO3) ≤0,005%
Calcium ≤0,05%
Potassium ≤0,0025%
Sodium ≤0,005%
Strontium ≤0,1%
Iron ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR