Bovine collagen, MP Biomedicals

Supplier: MP Biomedicals

0219349220
ICNA0219349220EA 1940 SGD
ICNA0219349220
Bovine collagen, MP Biomedicals
Proteins and Peptides

Collagen is a group of naturally occurring proteins found in animals, especially in the flesh and connective tissues of vertebrates. It is a component of skin, bone, tendon, and other fibrous connective tissues.


It is used to form actual culture gels, and human fibroblasts cultured in these gels are able to condense the collagen to a corium-like structure by cell contraction. A highly purified collagen solution suitable for 3-D matrix formation in cell culture.

Order Now

SPECIFICATIONS

Protein/peptide name Collagen
Environmentally Preferable
Species Bovine
Source
CAS No. 9007-34-5
Concentration 0,3% solution
Colour Colourless
Formulation Liquid
Storage conditions +4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR