Tritirachium album proteinase K, MP Biomedicals

Supplier: MP Biomedicals

0219350425 0219350483 0219350490 0219350491
ICNA0219350425EA 188 SGD
ICNA0219350425 ICNA0219350483 ICNA0219350490 ICNA0219350491
Tritirachium album proteinase K, MP Biomedicals
Enzymes

Suitable for both protein and nucleic acid isolation. Exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters. Also active with nitroanilides of amino acids with protected amino groups, excluding arginine. Useful in the isolation of DNA and RNA, in the analysis of membrane structures and protein structure. Proteolytic activity: >30mAnson Units/mg Unit definition: One Anson unit is the amount of enzyme which liberates 1 µmol of Folin-positive amino acids per minute at pH 7,5 and 35 °C, using hemoglobin as substrate.


  • Chromatographically purified


Protein content: >80%
RNase: <5×10⁻⁴ U/mg
DNase: <5×10⁻⁴ U/mg

Order Now

SPECIFICATIONS

Enzyme name Proteinase K
Environmentally Preferable
Source
Species Tritirachium album
CAS No. 39450-01-6
Molecular weight ~30 kDa
Form White/Off-white lyophilized powder
Storage conditions +4 °C
Enzyme activity >30 mAnson u/mg
pH 8,9

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR