Sodium pyruvate ≥99%, white powder cell culture reagent

Supplier: MP Biomedicals
Danger

0219473490 0219473425 0219473480
ICNA0219473490EA 1610 SGD
ICNA0219473490 ICNA0219473425 ICNA0219473480
Sodium pyruvate ≥99%, white powder cell culture reagent
Sodium pyruvate

Storage: +4 °C
Pyruvic acid is an intermediate in sugar metabolism and in enzymatic carbohydrate degradation (alcoholic fermentation) where it is converted to acetaldehyde and CO2 by carboxylase. In muscle, pyruvic acid (derived from glycogen) is reduced to lactic acid during exertion, which is reoxidized and partially retransformed to glycogen during rest. It improves coliform recovery when present in culture medium. It is involved in a metabolic regulatory pathway activated by mitochondrial oxidants. Pyruvate is involved in respiratory regulation in plants by interacting with alternative oxidase at a conserved cysteine residue. It may help prevent hydrogen peroxide mediated cell death.
Sodium pyruvate is utilized as a component in culture broth and media. Sodium pyruvate is used in Wallen fermentation medium to enhance the conversion of oleic acid to 10-ketostearic acid by Bacillus sphaericus. Sodium pyruvate has also been used to establish stably transfected human B cell lines.
Sodium Pyruvate has shown antioxidant properties and protective effects against oxygen radicals. Pyruvate is produced as part of glycolysis and is an intermediate in many metabolic pathways. It can be converted into acetyl CoA and enter the TCA Cycle.


  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Soluble in water (1 M [110 mg/mL] - clear, colorless solution).

Formula: CH₃COCOONa
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: >300 °C (lit.)
Density: 1,58 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
IR Conforms to Standard
pH (1% aq soln) 5-7

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR