Sodium chloride 99,5, powder_white Cell Culture

Supplier: MP Biomedicals

Synonyms: Saline, NaCl, Sea salt, Halite

02194738.5 0219473801 0219473805 0219473810
ICNA02194738.5EA 102 SGD
ICNA02194738.5 ICNA0219473801 ICNA0219473805 ICNA0219473810
Sodium chloride 99,5, powder_white Cell Culture
Sodium chloride

Storage: Store at Room Temperature (15-30 °C)
Sodium chloride is a commonly used chemical which is found widely in nature. It is considered to be an essential nutrient. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.
Sodium chloride is used in a wide variety of biochemical applications, including intravenous fluids (normal saline is 0.9% w/v in water), density gradients, a diluent to increase ionic strength in buffers or culture media and in salt-out procedures in the isolation of DNA. It has also been used in high concentrations for preservation of foods since bacteria cannot grow in high salt conditions. Sodium chloride is widely used in protein crystallization.


  • Cell Culture Reagent Grade
  • Vapor Pressure: 1 mm Hg at 865 °C (lit.)
  • Presentation: White Powder
  • Soluble in water (357 mg/mL at 25 °C; 384 mg/mL at 100 °C), glycerol (100 mg/mL); very slightly soluble in ethanol. Almost insoluble in concentrated HCl.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1461 °C (1013 hPa) (lit.)
Melting Pt: 801 °C (lit.)
Density: 2.17 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99.5%
pH (5% aq soln) 5.0-9.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR