Sodium chloride 99,5, powder_crystalline_white Molecular_Biology

Supplier: MP Biomedicals

Synonyms: Saline, NaCl, Sea salt, Halite

0219484810 02194848.5 0219484801 0219484805
ICNA0219484810EA 665 SGD
ICNA0219484810 ICNA02194848.5 ICNA0219484801 ICNA0219484805
Sodium chloride 99,5, powder_crystalline_white Molecular_Biology
Sodium chloride

Sodium chloride is a commonly used chemical which is found widely in nature. It is considered to be an essential nutrient. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.
Sodium chloride is used in a wide variety of biochemical applications, including intravenous fluids (normal saline is 0.9% w/v in water 10), density gradients 3,6, a diluent to increase ionic strength in buffers or culture media and in salt-out procedures in the isolation of DNA. It has also been used in high concentrations for preservation of foods since bacteria cannot grow in high salt conditions. A salt-ice mixture in the ratio of approximately 33 g sodium chloride to 100 g ice (at -1°C) will drop in temperature to as low as -21°C, depending on the rate of stirring and the size of the ice chunks.


  • Molecular Biology Reagent Grade
  • Vapor Pressure: 1 mm Hg at 865 °C (Lit.)
  • Presentation: White Crystalline Powder
  • Soluble in water (357 mg/mL at 25 °C; 384 mg/mL at 100 °C), glycerol (100 mg/mL); very slightly soluble in ethanol.
  • Can be sterilized by autoclaving or sterile filtering.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1461 °C (1013 hPa) (lit.)
Melting Pt: 801 °C (lit.)
Density: 2.17 g/cm³ (20 °C) (sat. 1.202 (25°C)) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99.5%
Barium Passes
Bromide ≤0.01%
Insoluble Matter ≤0.005%
Magnesium ≤0.001%
Sulfate ≤0.004%
Iron ≤0.0002%
Calcium ≤0.002%
Chlorates and Nitrates ≤0.003%
Phosphate ≤0.0005%
Heavy Metals ≤0.0005%
Potassium ≤0.005%
Iodide ≤0.002%
pH (5% aq soln, 25°C) 5.0 - 9.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR