Ammonium acetate 7.5 M, clear solution

Supplier: MP Biomedicals
Warning

Synonyms: Acetic acid ammonium salt

0219875991 0219875980
ICNA0219875991EA 899 SGD
ICNA0219875991 ICNA0219875980
Ammonium acetate 7.5 M, clear solution
Ammonium acetate

Ammonium acetate is a chemical compound which is derived from the reaction of ammonia and acetic acid. Ammonium acetate is volatile at low pressures. Because of which it has been used to replace cell buffers with non-volatile salts, in preparing samples for mass spectrometry. It is used as a buffer for mobile phases for HPLC with ELSD detection.


Ammonium acetate is a widely used reagent in molecular biology and chromatography. Its applications include the purification and precipitation of DNA and protein crystallization. Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides. Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.


Store at +4 °C.


Presentation: Clear Solution

Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Density: 1.17 (lit.)
Flash Pt: 136
Storage Temperature: Refrigerator
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Concentration 7.5 M aq (578.25 g/L)
Sterility Sterile


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR