Briggs and Salt Mixture, MP Biomedicals
Supplier: MP Biomedicals
0290283410
ICNA0290283410EA 1520 SGD
ICNA0290283410
Briggs and Salt Mixture, MP Biomedicals
Animal Supplies Animal Feed
This powdered salt mixture is used as animal diet.
It contains calcium carbonate, calcium phosphate, copper sulphate, ferric citrate, magnesium sulphate, manganese sulphate, potassium chloride, potassium iodate, sodium chloride, sodium phosphate dibasic and zinc carbonate.
Caution: Store at room temperature.