Hubbel, Mendel and Wakeman Salt Mixture, MP Biomedicals
Supplier: MP Biomedicals
0290283802
ICNA0290283802EA 193 SGD
ICNA0290283802
Hubbel, Mendel and Wakeman Salt Mixture, MP Biomedicals
Animal Supplies Animal Feed
Hubbel, Mendel and Wakeman powder does not contain zinc, is low in manganese and phosphorus, and has a high calcium to phosphorus ratio.
Designed for use in formulating animal research diets, especially with relatively high casein levels.
For complete composition see product datasheet.